(R)-α-(2-Oxiranyl)anisole 98%

Supplier: Ambeed
Danger

Synonyms: (R)-Glycidyl Phenyl ether, (R)-2-(Phenoxymethyl)oxirane, (R)-1,2-Epoxy-3-phenoxypropane

A667499-1G A667499-25G A667499-5G
76819-846EA 20.16 USD
76819-846 76819-848 76819-850
(R)-α-(2-Oxiranyl)anisole 98%
(R)-α-(2-Oxiranyl)anisole

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.1745 g/mol
MDL Number: MFCD06659025
CAS Number: 71031-02-2
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR