4-Amino-2-hydroxypyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A664891-100MG A664891-1G A664891-250MG
76819-822EA 65.78 USD
76819-822 76819-824 76819-826
4-Amino-2-hydroxypyrimidine-5-carbonitrile 95%
4-Amino-2-hydroxypyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₄O
MW: 136.11 g/mol
MDL Number: MFCD09863919
CAS Number: 16462-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR