Methyl 2-methylpyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A661893-100MG A661893-5G A661893-250MG A661893-1G
76819-802EA 63 USD
76819-802 76819-804 77673-804 77882-514
Methyl 2-methylpyrimidine-4-carboxylate 95%
Methyl 2-methylpyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD13174780
CAS Number: 73955-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR