2,6-Dimethylbenzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: m-Xylene-2-sulphonyl chloride, 2,6-Dimethylbenzene-1-sulphonyl chloride

A660051-100MG A660051-250MG
76819-798EA 120.95 USD
76819-798 76819-800
2,6-Dimethylbenzenesulfonyl chloride 95%
2,6-Dimethylbenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
MDL Number: MFCD03789123
CAS Number: 2905-29-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR