3-(2,4-Difluorobenzoyl)propionic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 4-(2,4-Difluorophenyl)-4-oxobutyric Acid, 4-(2,4-Difluorophenyl)-4-oxobutanoic acid

A658685-100G A658685-1G A658685-25G A658685-5G
76819-786EA 1689 USD
76819-786 76819-788 76819-790 76819-792
3-(2,4-Difluorobenzoyl)propionic acid 95+%
3-(2,4-Difluorobenzoyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₈F₂O₃
MDL Number: MFCD00143016
CAS Number: 110931-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR