2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-oxa-6-azaspiro[3.5]nonane-6-carboxylate

A657831-100MG A657831-250MG
76819-776EA 43 USD
76819-776 76819-778
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 98%
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3 g/mol
MDL Number: MFCD12545892
CAS Number: 1245816-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR