3-Bromo-6-chloro-7-azaindole 97%

Supplier: Ambeed
Warning

A655914-1G A655914-250MG A655914-5G
76819-764EA 35 USD
76819-764 76819-766 76819-768
3-Bromo-6-chloro-7-azaindole 97%
3-Bromo-6-chloro-7-azaindole

Precaución: for Research Use Only

Formula: C7H4BrClN2
MDL Number: MFCD12962923
CAS Number: 1190321-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR