2',4'-Dibromoacetophenone 95%

Supplier: Ambeed
Warning

A653805-250MG A653805-5G A653805-1G A653805-25G
76819-760EA 30.77 USD
76819-760 76819-762 77673-780 77673-782
2',4'-Dibromoacetophenone 95%
2',4'-Dibromoacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Br₂O
MW: 277.94 g/mol
MDL Number: MFCD00152057
CAS Number: 33243-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR