3,5-Dichloro-2-nitroaniline 95%

Supplier: Ambeed
Warning

A652723-1G A652723-250MG
76819-756EA 168.69 USD
76819-756 76819-758
3,5-Dichloro-2-nitroaniline 95%
3,5-Dichloro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MW: 207.014 g/mol
MDL Number: MFCD01463654
CAS Number: 122584-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR