2,6-Difluoro-3-hydroxypyridine 96%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-3-hydroxy-pyridine, 2,6-Difluoropyridin-3-ol

A648885-1G A648885-250MG
76819-732EA 96.55 USD
76819-732 77673-762
2,6-Difluoro-3-hydroxypyridine 96%
2,6-Difluoro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃F₂NO
MW: 131.08 g/mol
MDL Number: MFCD11044351
CAS Number: 209328-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR