4-Bromo-3-isopropylbenzoic acid ≥95%

Supplier: Ambeed
Warning

A647819-100MG A647819-1G
76819-730EA 31.83 USD
76819-730 77573-082
4-Bromo-3-isopropylbenzoic acid ≥95%
4-Bromo-3-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
MDL Number: MFCD15833351
CAS Number: 741698-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR