2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol 98%

Supplier: Ambeed
Warning

A643284-100G A643284-25G A643284-500G
76819-708EA 23.34 USD
76819-708 76819-710 76819-712
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol 98%
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol

Precaución: for Research Use Only

Formula: C₃₀H₂₉N₃O
MW: 447.58 g/mol
MDL Number: MFCD00134705
CAS Number: 70321-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR