Dipentaerythritol hexaacrylate 95%

Supplier: Ambeed
Warning

A642695-1000G A642695-100G A642695-25G A642695-500G
76819-700EA 47 USD
76819-700 76819-702 76819-704 76819-706
Dipentaerythritol hexaacrylate 95%
Dipentaerythritol hexaacrylate

Precaución: for Research Use Only

Formula: C₂₈H₃₄O₁₃
MDL Number: MFCD13194852
CAS Number: 29570-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR