2H-Pyrazolo[3,4-c]pyridine 95%

Supplier: Ambeed
Warning

A640911-1G A640911-250MG A640911-5G
76819-696EA 21 USD
76819-696 76819-698 77884-502
2H-Pyrazolo[3,4-c]pyridine 95%
2H-Pyrazolo[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD13175323
CAS Number: 271-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR