1-Boc-azetidine-3-acetaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-oxoethyl)azetidine-1-carboxylate

A639819-100MG A639819-1G A639819-250MG A639819-5G
76819-694EA 70.02 USD
76819-694 77573-064 77613-984 77673-734
1-Boc-azetidine-3-acetaldehyde 95%
1-Boc-azetidine-3-acetaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.247 g/mol
MDL Number: MFCD18374993
CAS Number: 152537-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR