Methyl 2-fluoro-5-hydroxy-4-methylbenzoate 98%

Supplier: Ambeed
Warning

A639088-100MG A639088-1G A639088-250MG A639088-5G
76819-684EA 24.41 USD
76819-684 76819-686 76819-688 76819-690
Methyl 2-fluoro-5-hydroxy-4-methylbenzoate 98%
Methyl 2-fluoro-5-hydroxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
MDL Number: MFCD19440272
CAS Number: 1378655-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR