Ethyl 2-bromo-2-cyclopropylacetate 95%

Supplier: Ambeed
Danger

A636646-1G A636646-250MG A636646-25G A636646-5G A636646-100MG
76819-676EA 35 USD
76819-676 76819-678 77673-730 77884-499 77884-498
Ethyl 2-bromo-2-cyclopropylacetate 95%
Ethyl 2-bromo-2-cyclopropylacetate

Precaución: for Research Use Only

Formula: C₇H₁₁BrO₂
MDL Number: MFCD18837719
CAS Number: 1200828-74-5
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR