(1R,2R)-Bis(4-methoxyphenyl)-1,2-ethanediamine 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-1,2-Di(4-methoxyphenyl)-1,2-diaminoethane

A635119-1G A635119-250MG A635119-100MG
76819-670EA 93.36 USD
76819-670 76819-672 77673-726
(1R,2R)-Bis(4-methoxyphenyl)-1,2-ethanediamine 97%
(1R,2R)-Bis(4-methoxyphenyl)-1,2-ethanediamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MW: 272.3422 g/mol
MDL Number: MFCD08460192
CAS Number: 58520-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR