7-Bromo-3-nitroquinolin-4-ol 97%

Supplier: Ambeed
Warning

A634095-100MG A634095-1G A634095-250MG A634095-5G
76819-652EA 8.49 USD
76819-652 76819-654 76819-656 76819-658
7-Bromo-3-nitroquinolin-4-ol 97%
7-Bromo-3-nitroquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O₃
MW: 269.05 g/mol
MDL Number: MFCD17170311
CAS Number: 723280-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR