6-Bromo-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98+%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-methylpyridine-3-boronic acid pinacol ester

A633967-1G A633967-250MG A633967-5G
76819-646EA 201.58 USD
76819-646 76819-648 76819-650
6-Bromo-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98+%
6-Bromo-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BBrNO₂
MW: 297.98 g/mol
MDL Number: MFCD13182151
CAS Number: 1451391-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR