4-Bromo-2-chloro-6-nitrophenol 98%

Supplier: Ambeed
Danger

A633712-1G A633712-25G A633712-5G
76819-640EA 14 USD
76819-640 76819-642 76819-644
4-Bromo-2-chloro-6-nitrophenol 98%
4-Bromo-2-chloro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₃
MDL Number: MFCD09835006
CAS Number: 58349-01-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR