3-(Difluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A632545-100MG A632545-1G A632545-250MG A632545-5G
76819-624EA 16 USD
76819-624 76819-626 77673-720 77882-465
3-(Difluoromethyl)benzoic acid 98%
3-(Difluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD17480442
CAS Number: 55805-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR