2-Iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Danger

A631368-100MG A631368-1G A631368-250MG
76819-612EA 5 USD
76819-612 76819-614 76819-616
2-Iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%
2-Iodo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉IN₂O₂S
MDL Number: MFCD11559008
CAS Number: 282734-63-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR