L(+)-Lysine monohydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl

A630759-100G A630759-25G A630759-500G A630759-1000G
76819-606EA 10 USD
76819-606 76819-608 76819-610 77673-710
L(+)-Lysine monohydrochloride 95%
L(+)-Lysine monohydrochloride

Precaución: for Research Use Only

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
MDL Number: MFCD00064564
CAS Number: 657-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR