5-Bromo-2-chloro-4-ethoxypyridine 95%

Supplier: Ambeed
Warning

A630400-100MG A630400-1G A630400-5G
76819-600EA 16.98 USD
76819-600 76819-602 76819-604
5-Bromo-2-chloro-4-ethoxypyridine 95%
5-Bromo-2-chloro-4-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MW: 236.49 g/mol
MDL Number: MFCD21603864
CAS Number: 52311-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR