Diethyl 1H-pyrrole-2,4-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
A627450-100G
A627450-10G
A627450-1G
A627450-250MG
A627450-25G
A627450-5G
76819-578EA
2522.83
USD
76819-578
76819-580
76819-582
76819-584
76819-586
76819-588
Diethyl 1H-pyrrole-2,4-dicarboxylate 95%
Diethyl 1H-pyrrole-2,4-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₄ MW: 211.21 g/mol |
MDL Number:
MFCD11977148 CAS Number: 55942-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...