1,7-Naphthyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A627102-1G A627102-250MG A627102-100MG
76819-574EA 802.05 USD
76819-574 76819-576 77573-036
1,7-Naphthyridine-3-carboxylic acid 97%
1,7-Naphthyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MW: 174.156 g/mol
MDL Number: MFCD11044184
CAS Number: 250674-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR