4-Methyl-1-piperazinesulfonyl Chloride 98%

Supplier: Ambeed
Danger

A625361-1G A625361-250MG A625361-5G
76819-562EA 45 USD
76819-562 77573-030 77673-692
4-Methyl-1-piperazinesulfonyl Chloride 98%
4-Methyl-1-piperazinesulfonyl Chloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O₂S
MDL Number: MFCD09731871
CAS Number: 1688-95-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR