3-Bromo-6-chloro-2-fluorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A624763-1G
A624763-250MG
A624763-5G
76819-550EA
28
USD
76819-550
76819-552
76819-554
3-Bromo-6-chloro-2-fluorobenzaldehyde 98%
3-Bromo-6-chloro-2-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃BrClFO MW: 237.4500 g/mol |
MDL Number:
MFCD11111905 CAS Number: 886615-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...