4-Chloro-1,2-dimethoxybenzene 98%

Supplier: Ambeed
Warning

A624516-250MG A624516-5G A624516-1G
76819-546EA 15 USD
76819-546 76819-548 77673-688
4-Chloro-1,2-dimethoxybenzene 98%
4-Chloro-1,2-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MDL Number: MFCD01741851
CAS Number: 16766-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR