6-Chloro-2-fluoro-3-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A623539-1G A623539-250MG A623539-5G
76819-534EA 40.32 USD
76819-534 76819-536 76819-538
6-Chloro-2-fluoro-3-methoxybenzaldehyde 95%
6-Chloro-2-fluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD04115946
CAS Number: 112641-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR