2-Methoxypyridine-3-sulfonyl chloride 98+%

Supplier: Ambeed
Danger

A622966-1G A622966-25G A622966-5G
76819-528EA 59.42 USD
76819-528 76819-530 76819-532
2-Methoxypyridine-3-sulfonyl chloride 98+%
2-Methoxypyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₆ClNO₃S
MW: 207.63 g/mol
MDL Number: MFCD13188795
CAS Number: 219715-44-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR