(4-Butoxy-3-fluorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

A622538-1G A622538-250MG A622538-5G
76819-522EA 7 USD
76819-522 76819-524 76819-526
(4-Butoxy-3-fluorophenyl)boronic acid 95%
(4-Butoxy-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
MDL Number: MFCD06411286
CAS Number: 156487-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR