3-(Dimethylamino)-1,2-propanediol 98%

Supplier: Ambeed
Danger

A621524-10G A621524-25G
76819-516EA 6 USD
76819-516 76819-518 76819-520
3-(Dimethylamino)-1,2-propanediol 98%
3-(Dimethylamino)-1,2-propanediol

Precaución: for Research Use Only

Formula: C5H13NO2
MDL Number: MFCD00004714
CAS Number: 623-57-4
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR