2,6-Di(benzo[d]oxazol-2-yl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Bis(benzo[d]oxazol-2-yl)pyridine

A617640-100MG A617640-1G A617640-250MG
76819-496EA 50 USD
76819-496 76819-498 76819-500
2,6-Di(benzo[d]oxazol-2-yl)pyridine 98%
2,6-Di(benzo[d]oxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₁₁N₃O₂
MDL Number: MFCD05241117
CAS Number: 33858-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR