3-(Bromomethyl)-2-cyclopropyl-4-(4-fluorophenyl)quinoline 95%

Supplier: Ambeed
Danger

A616621-1G A616621-250MG
76819-492EA 47.75 USD
76819-492 76819-494
3-(Bromomethyl)-2-cyclopropyl-4-(4-fluorophenyl)quinoline 95%
3-(Bromomethyl)-2-cyclopropyl-4-(4-fluorophenyl)quinoline

Precaución: for Research Use Only

Formula: C₁₉H₁₅BrFN
MW: 356.23 g/mol
MDL Number: MFCD09031391
CAS Number: 154057-56-4
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR