1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride 97%

Supplier: Ambeed
Warning

A616525-10G A616525-1G A616525-5G
76819-486EA 42 USD
76819-486 76819-488 76819-490
1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride 97%
1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂·2ClH
MW: 291.26 g/mol
MDL Number: MFCD09475546
CAS Number: 1228879-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR