6-Methoxyisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A611912-100MG A611912-250MG A611912-5G A611912-1G
76819-472EA 18 USD
76819-472 76819-474 77613-904 77673-654
6-Methoxyisoquinolin-1(2H)-one 95%
6-Methoxyisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD12405870
CAS Number: 26829-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR