tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A610195-100G A610195-10G A610195-1G A610195-250MG A610195-5G A610195-25G
76819-452EA 396 USD
76819-452 76819-454 76819-456 76819-458 76819-460 77884-469
tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate 98%
tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₃H₃₂N₂O₃
MW: 384.5118 g/mol
MDL Number: MFCD09833420
CAS Number: 144163-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR