1-Benzyl-4-cyano-4-phenylpiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-4-phenylpiperidine-4-carbonitrile

A608459-1G A608459-5G
76819-448EA 11.67 USD
76819-448 76819-450
1-Benzyl-4-cyano-4-phenylpiperidine 98%
1-Benzyl-4-cyano-4-phenylpiperidine

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₂
MW: 276.38 g/mol
MDL Number: MFCD00044826
CAS Number: 56243-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR