5-Aminopyrimidine-4,6-diol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-4,6-dihydroxypyrimidine

A605401-1G A605401-250MG A605401-25G
76819-438EA 49.87 USD
76819-438 76819-440 76819-442
5-Aminopyrimidine-4,6-diol 98%
5-Aminopyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MW: 127.101 g/mol
MDL Number: MFCD00239457
CAS Number: 69340-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR