2-Bromo-3,4,5-trifluoro-1-nitrobenzene 98%

Supplier: Ambeed
Warning

A604855-10G A604855-1G A604855-25G A604855-5G
76819-430EA 169.75 USD
76819-430 76819-432 76819-434 76819-436
2-Bromo-3,4,5-trifluoro-1-nitrobenzene 98%
2-Bromo-3,4,5-trifluoro-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₆HBrF₃NO₂
MW: 255.97 g/mol
MDL Number: MFCD09955450
CAS Number: 1020718-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR