6-Methoxybenzo[b]thiophene-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxybenzo[b]thiophene-2-carbaldehyde

A603317-100MG A603317-1G A603317-250MG
76819-416EA 46.68 USD
76819-416 76819-418 76819-420
6-Methoxybenzo[b]thiophene-2-carboxaldehyde 98%
6-Methoxybenzo[b]thiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MW: 192.234 g/mol
MDL Number: MFCD11101038
CAS Number: 1001203-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR