4-(trans-2-Carboxyvinyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

A602573-1G A602573-250MG A602573-5G A602573-25G
76819-410EA 55 USD
76819-410 76819-412 76819-414 77884-450
4-(trans-2-Carboxyvinyl)phenylboronic acid 96%
4-(trans-2-Carboxyvinyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₉BO₄
MDL Number: MFCD01075704
CAS Number: 159896-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR