tert-Butyl ((1R,3R)-3-hydroxycyclopentyl)carbamate 95%

Supplier: Ambeed
Warning

A601617-100MG A601617-1G A601617-250MG
76819-398EA 94.43 USD
76819-398 76819-400 76819-402
tert-Butyl ((1R,3R)-3-hydroxycyclopentyl)carbamate 95%
tert-Butyl ((1R,3R)-3-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD19442807
CAS Number: 1290191-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR