1'-Benzylspiro[indoline-3,4'-piperidine] 95%

Supplier: Ambeed
Warning

A598653-100MG A598653-250MG
76819-386EA 76.39 USD
76819-386 76819-388
1'-Benzylspiro[indoline-3,4'-piperidine] 95%
1'-Benzylspiro[indoline-3,4'-piperidine]

Precaución: for Research Use Only

Formula: C₁₉H₂₂N₂
MW: 278.3900 g/mol
MDL Number: MFCD04108106
CAS Number: 474538-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR