Methyl benzo[b]thiophene-4-carboxylate 95%

Supplier: Ambeed
Warning

A598128-250MG A598128-1G A598128-100MG
76819-384EA 41 USD
76819-384 77613-860 77884-442
Methyl benzo[b]thiophene-4-carboxylate 95%
Methyl benzo[b]thiophene-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MDL Number: MFCD20527696
CAS Number: 100590-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR