Methyl 5-bromo-2-(trifluoromethoxy)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-(trifluoromethoxy)benzoic acid methyl ester

A595050-1G A595050-250MG
76819-350EA 148.53 USD
76819-350 76819-352
Methyl 5-bromo-2-(trifluoromethoxy)benzoate 98%
Methyl 5-bromo-2-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₃
MW: 299.041 g/mol
MDL Number: MFCD06203799
CAS Number: 773874-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR