6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A594287-100MG
A594287-1G
76819-346EA
21.22
USD
76819-346
76819-348
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carboxylic acid 95%
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₈O₃S MW: 184.212 g/mol |
MDL Number:
MFCD12088019 CAS Number: 933747-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...