Di-tert-butyl chloromethyl phosphate 95%
Supplier: Ambeed
|
Warning
|
A593073-100G
A593073-10G
A593073-1G
A593073-25G
A593073-5G
76819-332EA
293
USD
76819-332
76819-334
76819-336
76819-338
76819-340
Di-tert-butyl chloromethyl phosphate 95%
Di-tert-butyl chloromethyl phosphate
Precaución: for Research Use Only
Formula:
C₉H₂₀ClO₄P |
MDL Number:
MFCD12165895 CAS Number: 229625-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...